Assay Details
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083773.1 | NP_001077242.1 | 1 - 2 | 111 | 88 | |
GenBank mRNA | BC122804.1 | - | 1 - 2 | 111 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122804.1, NM_001083773.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
