Assay Details
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175779.3 | NP_786973.1 | 1 - 2 | 154 | 147 | |
GenBank mRNA | BC120081.1 | - | 1 - 2 | 154 | 147 | |
BT029887.1 | - | 1 - 2 | 119 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120081.1, NM_175779.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
