Assay Details
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175779.3 | NP_786973.1 | 4 - 5 | 619 | 65 | |
GenBank mRNA | BC120081.1 | - | 4 - 5 | 619 | 65 | |
BT029887.1 | - | 4 - 5 | 584 | 65 | ||
M17025.1 | - | 4 - 5 | 499 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
