Assay Details
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175779.3 | NP_786973.1 | 5 - 6 | 744 | 109 | |
GenBank mRNA | BC120081.1 | - | 5 - 6 | 744 | 109 | |
BT029887.1 | - | 5 - 6 | 709 | 109 | ||
M17025.1 | - | 5 - 6 | 624 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
