Assay Details
Gene Symbol: LTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013401.2 | NP_001013419.1 | 1 - 2 | 113 | 118 | |
GenBank mRNA | BC114670.1 | - | 1 - 2 | 113 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
