Assay Details
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014845.3 | NP_001014845.1 | 2 - 3 | 498 | 97 | |
GenBank mRNA | BC133432.1 | - | 2 - 3 | 499 | 97 | |
EU935419.1 | - | 2 - 3 | 293 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
