Assay Details
Gene Symbol: PLSCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046053.2 | NP_001039518.1 | 4 - 5 | 966 | 98 | |
GenBank mRNA | BC114676.1 | - | 4 - 5 | 614 | 98 | |
BT029864.1 | - | 4 - 5 | 966 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
