Assay Details
Gene Symbol: NFE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077899.2 | NP_001071367.2 | 3 - 4 | 1054 | 152 | |
GenBank mRNA | BC126669.1 | - | 2 - 3 | 295 | 152 | |
BC134642.1 | - | 3 - 4 | 1054 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126669.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
