Assay Details
Gene Symbol: PELI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078004.1 | NP_001071472.1 | 5 - 6 | 895 | 83 | |
GenBank mRNA | BC126542.1 | - | 5 - 6 | 895 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
