Assay Details
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078079.1 | NP_001071547.1 | 4 - 5 | 305 | 107 | |
GenBank mRNA | BC126840.1 | - | 4 - 5 | 305 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
