Assay Details
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079794.1 | NP_001073262.1 | 1 - 2 | 146 | 133 | |
GenBank mRNA | BC123791.1 | - | 1 - 2 | 164 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
