Assay Details
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080221.2 | NP_001073690.1 | 3 - 4 | 298 | 63 | |
GenBank mRNA | AB098954.1 | - | 3 - 4 | 298 | 63 | |
BC126786.1 | - | 3 - 4 | 299 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
