Assay Details
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080242.2 | NP_001073711.1 | 6 - 7 | 639 | 58 | |
GenBank mRNA | BC133594.1 | - | 6 - 7 | 639 | 58 | |
DQ355511.1 | - | 5 - 6 | 561 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
