Assay Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080253.1 | NP_001073722.1 | 3 - 4 | 515 | 75 | |
GenBank mRNA | BC126633.1 | - | 3 - 4 | 515 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
