Assay Details
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080265.1 | NP_001073734.1 | 2 - 3 | 176 | 67 | |
GenBank mRNA | BC126727.1 | - | 2 - 3 | 176 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
