Assay Details
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080268.2 | NP_001073737.1 | 6 - 7 | 516 | 94 | |
GenBank mRNA | BC126638.1 | - | 6 - 7 | 517 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
