Assay Details
Gene Symbol: MEPCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080283.2 | NP_001073752.2 | 2 - 3 | 1972 | 136 | |
GenBank mRNA | BT029808.1 | - | 2 - 3 | 1201 | 136 | |
EE245239.1 | - | 2 - 3 | 584 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
