Assay Details
Gene Symbol: WASF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081511.1 | NP_001074980.1 | 5 - 6 | 686 | 87 | |
GenBank mRNA | BC133284.1 | - | 5 - 6 | 686 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
