Assay Details
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081526.1 | NP_001074995.1 | 3 - 4 | 598 | 105 | |
GenBank mRNA | BC133335.1 | - | 3 - 4 | 598 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
