Assay Details
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081617.1 | NP_001075086.1 | 2 - 3 | 239 | 115 | |
GenBank mRNA | BC133396.1 | - | 2 - 3 | 239 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
