Assay Details
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081618.1 | NP_001075087.1 | 1 - 2 | 444 | 117 | |
GenBank mRNA | BC133414.1 | - | 1 - 2 | 444 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
