Assay Details
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082434.2 | NP_001075903.1 | 2 - 3 | 617 | 66 | |
GenBank mRNA | BC133569.1 | - | 2 - 3 | 618 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
