Assay Details
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082434.2 | NP_001075903.1 | 3 - 4 | 769 | 58 | |
GenBank mRNA | BC133569.1 | - | 3 - 4 | 770 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
