Assay Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082437.2 | NP_001075906.1 | 1 - 2 | 404 | 80 | |
GenBank mRNA | BC133634.1 | - | 1 - 2 | 405 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
