Assay Details
Gene Symbol: IGSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082439.1 | NP_001075908.1 | 4 - 5 | 1397 | 122 | |
GenBank mRNA | BC133513.1 | - | 4 - 5 | 1397 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
