Assay Details
Gene Symbol: LYPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082462.2 | NP_001075931.1 | 2 - 3 | 168 | 73 | |
GenBank mRNA | BC133550.1 | - | 2 - 3 | 170 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
