Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082613.1 | NP_001076082.1 | 3 - 4 | 3124 | 72 | |
GenBank mRNA | BC133353.1 | - | 3 - 4 | 3124 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
