Assay Details
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082614.1 | NP_001076083.1 | 1 - 2 | 121 | 74 | |
GenBank mRNA | BC133282.1 | - | 1 - 2 | 121 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
