Assay Details
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083373.1 | NP_001076842.1 | 2 - 3 | 697 | 80 | |
GenBank mRNA | BC114754.1 | - | 2 - 3 | 697 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
