Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083409.1 | NP_001076878.1 | 2 - 3 | 298 | 81 | |
GenBank mRNA | BC133322.1 | - | 2 - 3 | 298 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
