Assay Details
Gene Symbol: LAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083418.1 | NP_001076887.1 | 4 - 5 | 494 | 67 | |
GenBank mRNA | BC123658.1 | - | 4 - 5 | 494 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
