Assay Details
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083494.1 | NP_001076963.1 | 4 - 5 | 480 | 86 | |
GenBank mRNA | BC123884.1 | - | 4 - 5 | 480 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
