Assay Details
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083506.1 | NP_001076975.1 | 2 - 3 | 459 | 98 | |
GenBank mRNA | AB510901.1 | - | 2 - 3 | 481 | 98 | |
BC123854.1 | - | 2 - 3 | 459 | 98 | ||
LC271172.1 | - | 2 - 3 | 437 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
