Assay Details
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083511.1 | NP_001076980.1 | 5 - 6 | 743 | 102 | |
GenBank mRNA | BC123644.1 | - | 5 - 6 | 743 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
