Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083662.2 | NP_001077131.1 | 11 - 12 | 1510 | 125 | |
GenBank mRNA | BC123423.1 | - | 6 - 7 | 599 | 125 | |
BC134465.1 | - | 11 - 12 | 1511 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
