Assay Details
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083669.1 | NP_001077138.1 | 10 - 11 | 1469 | 63 | |
GenBank mRNA | BC146204.1 | - | 10 - 11 | 1559 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
