Assay Details
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083669.1 | NP_001077138.1 | 7 - 8 | 1205 | 74 | |
GenBank mRNA | BC146204.1 | - | 7 - 8 | 1295 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
