Assay Details
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083743.1 | NP_001077212.1 | 4 - 5 | 1293 | 95 | |
GenBank mRNA | BC134551.1 | - | 4 - 5 | 1293 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
