Assay Details
Gene Symbol: PTOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083780.1 | NP_001077249.1 | 2 - 3 | 465 | 83 | |
GenBank mRNA | BC134519.1 | - | 2 - 3 | 465 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
