Assay Details
Gene Symbol: TMEM182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083784.1 | NP_001077253.1 | 1 - 2 | 275 | 122 | |
GenBank mRNA | BC134437.1 | - | 1 - 2 | 275 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
