Assay Details
Gene Symbol: TMEM182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083784.1 | NP_001077253.1 | 2 - 3 | 376 | 123 | |
GenBank mRNA | BC134437.1 | - | 2 - 3 | 376 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
