Assay Details
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083791.1 | NP_001077260.1 | 2 - 3 | 388 | 96 | |
GenBank mRNA | BC134511.1 | - | 2 - 3 | 388 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
