Assay Details
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098859.2 | NP_001092329.1 | 3 - 4 | 633 | 98 | |
GenBank mRNA | AB035802.1 | - | 2 - 3 | 114 | 98 | |
BC146056.1 | - | 3 - 4 | 635 | 98 | ||
JN380921.1 | - | 3 - 4 | 444 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
