Assay Details
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105615.1 | NP_001099085.1 | 6 - 7 | 658 | 92 | |
GenBank mRNA | BC134699.1 | - | 6 - 7 | 658 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
