Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173966.3 | NP_776391.2 | 3 - 4 | 472 | 69 | |
GenBank mRNA | AF348421.1 | - | 3 - 4 | 279 | 69 | |
BC134755.1 | - | 3 - 4 | 473 | 69 | ||
EU276079.1 | - | 3 - 4 | 314 | 69 | ||
JN635499.1 | - | 3 - 4 | 310 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
