Assay Details
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174046.4 | NP_776471.3 | 6 - 7 | 1135 | 99 | |
NM_177956.3 | NP_808871.3 | 7 - 8 | 1222 | 99 | ||
NM_177957.3 | NP_808872.2 | 5 - 6 | 721 | 99 | ||
NM_177958.3 | NP_808873.2 | 6 - 7 | 808 | 99 | ||
GenBank mRNA | AF489573.1 | - | 6 - 7 | 930 | 99 | |
AF489574.1 | - | 7 - 8 | 1017 | 99 | ||
AF489575.1 | - | 5 - 6 | 723 | 99 | ||
AF489576.1 | - | 6 - 7 | 810 | 99 | ||
U27341.1 | - | 6 - 7 | 930 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
