Assay Details
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174448.5 | NP_776873.1 | 2 - 3 | 205 | 133 | |
GenBank mRNA | BC134566.1 | - | 2 - 3 | 135 | 133 | |
CN789028.1 | - | 2 - 3 | 205 | 133 | ||
J04196.1 | - | 1 - 2 | 58 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
