Assay Details
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174603.3 | NP_777028.2 | 2 - 3 | 225 | 75 | |
GenBank mRNA | AY033938.1 | - | 2 - 3 | 191 | 75 | |
BC133291.1 | - | 2 - 3 | 225 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
