Assay Details
Gene Symbol: JOSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014898.2 | NP_001014898.1 | 3 - 4 | 1112 | 74 | |
GenBank mRNA | BC140477.1 | - | 3 - 4 | 1112 | 74 | |
BT020624.1 | - | 3 - 4 | 826 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
