Assay Details
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031759.2 | NP_001026929.2 | 2 - 3 | 454 | 114 | |
GenBank mRNA | BC123427.1 | - | 2 - 3 | 454 | 114 | |
BT021628.1 | - | 2 - 3 | 785 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
